Learn More
Medchemexpress LLC 3,8-Diamino-6-phenylphenanthridine | 52009-64-0 | 285.34 | 500 MG
SDP
Catalog No. 5000453490
Encompass_Preferred
3,8-Diamino-6-phenylphenanthridine can bind to DNA by means of a non-specific binding along the DNA exterior. It is sold for research purposes only and not for patient use.
  • Binds to DNA via non-specific binding along the DNA exterior.
  • Available in various quantities.
  • Ships at room temperature in the continental US; storage at 4°C, protected from light.
  • In solvent, can be stored at -80°C for 6 months or -20°C for 1 month (protect from light).
Catalog No. 50-004-53490 Supplier Medchemexpress LLC Supplier No. HYW011511500MG
3,8-Diamino-6-phenylphenanthridine can bind to DNA by means of a non-specific binding along the DNA exterior. It is sold for research purposes only and not for patient use.
  • Binds to DNA via non-specific binding along the DNA exterior.
  • Available in various quantities.
  • Ships at room temperature in the continental US; storage at 4°C, protected from light.
  • In solvent, can be stored at -80°C for 6 months or -20°C for 1 month (protect from light).