Learn More
Medchemexpress LLC 3,8-Diamino-6-phenylphenanthridine | 52009-64-0 | MFCD00075152 | ≥98% | 285.34 | 100 MG
SDP
Catalog No. 5000454949
Encompass_Preferred
3,8-Diamino-6-phenylphenanthridine is a crystalline yellow solid that can bind to DNA by means of a non-specific binding along the DNA exterior. It has been investigated for its anti-tumor and anti-viral properties.
  • Used in the synthesis of rigid polyamides.
  • Investigated for its anti-tumor and anti-viral properties.
  • Binds to DNA by means of a non-specific binding along the DNA exterior.
Catalog No. 50-004-54949 Supplier Medchemexpress LLC Supplier No. HYW011511100MG
3,8-Diamino-6-phenylphenanthridine is a crystalline yellow solid that can bind to DNA by means of a non-specific binding along the DNA exterior. It has been investigated for its anti-tumor and anti-viral properties.
  • Used in the synthesis of rigid polyamides.
  • Investigated for its anti-tumor and anti-viral properties.
  • Binds to DNA by means of a non-specific binding along the DNA exterior.