Learn More
MSC CDI 2502MRMH 3/8" Drive Micrometer Torque Wrench 30 to 250 In/Lb, 1 In/Lb Graduation, 11-1/4" Overall Length, Pear Head
Catalog No. 501298035
Encompass
CDI 2502MRMH 3/8" Drive Micrometer Torque Wrench 30 to 250 In/Lb, 1 In/Lb Graduation, 11-1/4" Overall Length, Pear Head
Catalog No. 50-129-8035 Supplier MSC Supplier No. 87158051
CDI 2502MRMH 3/8" Drive Micrometer Torque Wrench 30 to 250 In/Lb, 1 In/Lb Graduation, 11-1/4" Overall Length, Pear Head