Learn More
MSC Proto Flex Handle, 3/8 Drive, 8.5 in, Chrome, 1 ea
Catalog No. 501920455
Encompass
This flex handle is designed to provide leverage and access in various applications. It features a head that flexes 90 degrees from horizontal, is constructed from heat-treated alloy steel, and has a chrome-plated finish for durability.
  • Head flexes 90 degrees from horizontal
  • Made of heat-treated alloy steel
  • Corrosion-resistant chrome-plated finish
  • Knurled grip style for secure handling
  • Can be used with 3/8 in drive sockets
  • Saves wear and tear on fine-tooth ratchets
  • Allows for precise angular displacement
Catalog No. 50-192-0455 Supplier MSC Supplier No. 38296695
This flex handle is designed to provide leverage and access in various applications. It features a head that flexes 90 degrees from horizontal, is constructed from heat-treated alloy steel, and has a chrome-plated finish for durability.
  • Head flexes 90 degrees from horizontal
  • Made of heat-treated alloy steel
  • Corrosion-resistant chrome-plated finish
  • Knurled grip style for secure handling
  • Can be used with 3/8 in drive sockets
  • Saves wear and tear on fine-tooth ratchets
  • Allows for precise angular displacement