Learn More
Chemglass Life Sciences Graded Seal, 3/8in OD Kovar to Borosilicate
SDP
Catalog No. CG30605
Encompass_Preferred
Graded Seal, 3/8in OD Kovar to Borosilicate. For connection of borosilicate glass components to metal. Kovar length is approximately 25mm on all sizes, while the borosilicate tubing is approximately 100mm.
Catalog No. CG30605 Supplier Chemglass Life Sciences Supplier No. CG30605
Graded Seal, 3/8in OD Kovar to Borosilicate. For connection of borosilicate glass components to metal. Kovar length is approximately 25mm on all sizes, while the borosilicate tubing is approximately 100mm.