Learn More
US Plastic Corporation 3/8" HOSE BARB POLYPROPYLENE T
SDP
Catalog No. NC1789520
Encompass
3/8" Hose Barb Polypropylene Three-way Ball Valve with EPDM seals & Red Handle
Catalog No. NC1789520 Supplier US Plastic Corporation Supplier No. 22363
Add to Cart
Edge
Add to Cart
3/8" Hose Barb Polypropylene Three-way Ball Valve with EPDM seals & Red Handle