Learn More
Ace Glass, Inc. Vacuum tubing, unreinforced red rubber, 3/8in ID x 3/8inwall weight, 1Ft
SDP
Catalog No. 50977642
Encompass
3/8 I.D. x 3/8in. wall weight hand-made vacuum tubing is manufactured from reinforced red rubber and features a heavy wall and low vapor pressure.
Catalog No. 50-977-642 Supplier Ace Glass, Inc. Supplier No. 1269010
3/8 I.D. x 3/8in. wall weight hand-made vacuum tubing is manufactured from reinforced red rubber and features a heavy wall and low vapor pressure.