Learn More
Analytical Sales & Services 3.8µL Mobile Phase Pre-Column Pre-Heater
Catalog No. 5000805669
Encompass
Advantage 3.8µL Mobile Phase Pre-Column Pre-Heater 0.005" I.D. x 30cm Stainless Steel 15.210 µL Internal Volume With (1) UHPLC Fitting
Qty/Pkg: EACH
Catalog No. 50-008-05669 Supplier Analytical Sales & Services Supplier No. ADV9500UHP
Advantage 3.8µL Mobile Phase Pre-Column Pre-Heater 0.005" I.D. x 30cm Stainless Steel 15.210 µL Internal Volume With (1) UHPLC Fitting
Qty/Pkg: EACH