Learn More
US Plastic Corporation 1/4" ID x 3/8" OD x 0.062" Wall Natural Polypropylene Tubing
SDP
Catalog No. NC1966850
Encompass
Polypropylene is ideal for transferring hot liquids or gases, or where higher temperatures and pressures are expected. It has many uses in manufacturing operations where extreme resistance to acids and alkalis is required.
Catalog No. NC1966850 Supplier US Plastic Corporation Supplier No. 58022
Polypropylene is ideal for transferring hot liquids or gases, or where higher temperatures and pressures are expected. It has many uses in manufacturing operations where extreme resistance to acids and alkalis is required.