Learn More
COLDER PRODUCTS COMPANY CPC 3/8 PTF valved Panel Mount Coupling Body Acetal Maximum Pressure Vacuum to 120 psi 8 3 bar Thread Size 3/8 in
Catalog No. 503131437
Encompass_Preferred
CPC 3/8 PTF valved Panel Mount Coupling Body Acetal Maximum Pressure Vacuum to 120 psi 8 3 bar Thread Size 3/8 in
Catalog No. 50-313-1437 Supplier COLDER PRODUCTS COMPANY Supplier No. PLCD1200612
CPC 3/8 PTF valved Panel Mount Coupling Body Acetal Maximum Pressure Vacuum to 120 psi 8 3 bar Thread Size 3/8 in