Learn More
Chemglass Life Sciences Cajon Flexible Tubing Only, 3/8" Size X 6" Length, Airfree, Schlenk
SDP
Catalog No. AF006904
Encompass_Preferred
Cajon Flexible Tubing Only, 3/8" Size X 6" Length, Airfree, Schlenk. Cajon Stainless Steel #321 Flexible Tubing is compressible by at least 20%, and extendable by 50% of its nominal listed length. The 1/4" and 3/8" sizes have a cuff length of 0.75" while the 1" size cuff length is 1".PLEASE NOTE: SINCE FLEXIBLE TUBING WILL CONTRACT WHEN EVACUATED, GLASS APPARATUS SHOULD NOT BE FIRMLY CLAMPED IN PLACE UNTIL VACUUM IS ESTABLISHED WITHIN THE SYSTEM
Catalog No. AF006904 Supplier Chemglass Life Sciences Supplier No. AF006904
Cajon Flexible Tubing Only, 3/8" Size X 6" Length, Airfree, Schlenk. Cajon Stainless Steel #321 Flexible Tubing is compressible by at least 20%, and extendable by 50% of its nominal listed length. The 1/4" and 3/8" sizes have a cuff length of 0.75" while the 1" size cuff length is 1".PLEASE NOTE: SINCE FLEXIBLE TUBING WILL CONTRACT WHEN EVACUATED, GLASS APPARATUS SHOULD NOT BE FIRMLY CLAMPED IN PLACE UNTIL VACUUM IS ESTABLISHED WITHIN THE SYSTEM