Learn More
Accuflow Systems 3/8ID X 1/2OD FEP 50FT/ROLL
Catalog No. NC0984185
Encompass_Preferred
*Chemically inert *Documented biocompatibility *Pharmafluor is fully characterized to all current standards *Non-cytotoxic, non-pyrogenic and non-hemolytic *Extremely low sorption characteristics compared to other flexible tubings *Fully autoclavable and sterilizable *Excellent barrier with very low permeability
Catalog No. NC0984185 Supplier Accuflow Systems Supplier No. PHFE6050006250
*Chemically inert *Documented biocompatibility *Pharmafluor is fully characterized to all current standards *Non-cytotoxic, non-pyrogenic and non-hemolytic *Extremely low sorption characteristics compared to other flexible tubings *Fully autoclavable and sterilizable *Excellent barrier with very low permeability