Learn More
MSC Value Collection Machine Screw, 8-32, 3/8 in, Pan, 100/Pk.
Catalog No. NC1895608
Encompass
This machine screw features an 8-32 thread, a 3/8 in length under head, and a pan head with a Phillips drive. It is designed for various fastening applications requiring corrosion resistance and a secure fit.
  • Constructed from 18-8 stainless steel for durability
  • Bright/uncoated finish for a clean appearance
  • Features 8-32 UNC thread for standard compatibility
  • Pan head design for a low-profile finish
  • Phillips drive for easy installation with common tools
  • Measures 3/8 in length under head
  • Without washer style
  • Made in China
Catalog No. NC1895608 Supplier MSC Supplier No. 67571489
This machine screw features an 8-32 thread, a 3/8 in length under head, and a pan head with a Phillips drive. It is designed for various fastening applications requiring corrosion resistance and a secure fit.
  • Constructed from 18-8 stainless steel for durability
  • Bright/uncoated finish for a clean appearance
  • Features 8-32 UNC thread for standard compatibility
  • Pan head design for a low-profile finish
  • Phillips drive for easy installation with common tools
  • Measures 3/8 in length under head
  • Without washer style
  • Made in China