Learn More
Malvern Panalytical 3/8IN SAMPLE TUBE
Catalog No. NC9569474
Encompass
This item has a minimum qty of 3 per supplier requirements.
SAMPLE TUBE, 3/8-in. (9.6-mm) OD x 6.1 in. (155-mm) long, approx 2 cm3 sample capacity
Catalog No. NC9569474 Supplier Malvern Panalytical Supplier No. 236/61703/00
This item has a minimum qty of 3 per supplier requirements.
SAMPLE TUBE, 3/8-in. (9.6-mm) OD x 6.1 in. (155-mm) long, approx 2 cm3 sample capacity