Learn More
Andwin Scientific Tough-Spots, 5-up, 3/8inch, Assorted, 5000/roll
SDP
Catalog No. NC1660917
Encompass_Preferred
Tough Spots on a roll are easy to write PVC cryogenic labels that accept writing from most lab markers. Tough Spots come in a variety of colors and sizes to ensure the right label for the right job!
Catalog No. NC1660917 Supplier Andwin Scientific Supplier No. SPRL1100
Tough Spots on a roll are easy to write PVC cryogenic labels that accept writing from most lab markers. Tough Spots come in a variety of colors and sizes to ensure the right label for the right job!