Learn More
Teknova 3.8M Ammonium Sulfate. 2L, Sterile 8 Pack.
SDP
Catalog No. 502387851
Encompass
Ammonium sulfate is a powerful inorganic disinfectant tbat can be used as a precipitating agent for both proteins and plasmids.
Catalog No. 50-238-7851 Supplier Teknova Supplier No. A05258PK
Ammonium sulfate is a powerful inorganic disinfectant tbat can be used as a precipitating agent for both proteins and plasmids.