Learn More
Ace Glass, Inc. PTFE 3/8in tube compression fitting to 1/2in male NPT
SDP
Catalog No. 50983400
Encompass
3/8 PTFE tube compression fitting to 1/2 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-983-400 Supplier Ace Glass, Inc. Supplier No. 1270946
3/8 PTFE tube compression fitting to 1/2 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.