Learn More
Ace Glass, Inc. PTFE 3/8in tube compression fitting to 3/8in male NPT
SDP
Catalog No. 50977681
Encompass
3/8 PTFE tube compression fitting to 3/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-977-681 Supplier Ace Glass, Inc. Supplier No. 1270936
3/8 PTFE tube compression fitting to 3/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.