Learn More
Ace Glass, Inc. PTFE 3/8" Tube x 1/4" Tube Compression Reducing Union
SDP
Catalog No. 50983406
Encompass
3/8 Tube x 1/4 Tube PTFE tube compression reducing union with body and ferrules made of virgin PTFE. The nut and gripper are made of PVDF. Performance at ambient room conditions: 120psig for 1/16 fittings linear, decreasing to 80psig for fittings, and 60psig for fittings larger than 3/4. Performance at elevated temperatures up to 85C/185F: 90psig for 1/16 fittings linear, decreasing to 60psig for 3/4 fittings, and 40psig for fittings larger than 3/4.
Catalog No. 50-983-406 Supplier Ace Glass, Inc. Supplier No. 1271112
3/8 Tube x 1/4 Tube PTFE tube compression reducing union with body and ferrules made of virgin PTFE. The nut and gripper are made of PVDF. Performance at ambient room conditions: 120psig for 1/16 fittings linear, decreasing to 80psig for fittings, and 60psig for fittings larger than 3/4. Performance at elevated temperatures up to 85C/185F: 90psig for 1/16 fittings linear, decreasing to 60psig for 3/4 fittings, and 40psig for fittings larger than 3/4.