Learn More
AVANTOR MASTERFLEX (VWR LLC) Barbed fittings, Reducing Connector, Natural PP, 3/8"" x 5/16"" ID,3/16"", 2-1/4""
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4061009/DEL
Encompass
Barbed fittings, Reducing Connector, Natural PP, 3/8" x 5/16" ID,3/16", 2-1/4", Ideal for low-pressure applicationsQuick and easy assembly without toolsChemical resistantAvailable in a variety of sizes, materials, and configurationsTemperture range 9 to 180¿F (-13 to 66¿C)Tubing ID 3/8" x 5/16"
Catalog No. NC1503231