Learn More
MSC Klein Tools 602-8-INS 3/8" Blade Width, 13-3/8" Overall Length Cabinet Slotted Screwdriver 8" Blade Length, Round Shank, Acetate with Rubber Grip Handle
Catalog No. 501298077
Encompass
Klein Tools 602-8-INS 3/8" Blade Width, 13-3/8" Overall Length Cabinet Slotted Screwdriver 8" Blade Length, Round Shank, Acetate with Rubber Grip Handle
Catalog No. 50-129-8077 Supplier MSC Supplier No. 52527918
Klein Tools 602-8-INS 3/8" Blade Width, 13-3/8" Overall Length Cabinet Slotted Screwdriver 8" Blade Length, Round Shank, Acetate with Rubber Grip Handle