Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,9-Bis(3-aminopropyl)-2,4,8,10-tetraoxaspiro[5.5]undecane, TCI America™
SDP

Catalog No. B172225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B172225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B172225G Supplier TCI America Supplier No. B172225G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21587-74-6
Molecular Formula C13H26N2O4
Molecular Weight (g/mol) 274.361
MDL Number MFCD00023818
InChI Key ANOPCGQVRXJHHD-UHFFFAOYSA-N
Synonym ATU
PubChem CID 88957
IUPAC Name 3-[3-(3-aminopropyl)-2,4,8,10-tetraoxaspiro[5.5]undecan-9-yl]propan-1-amine
SMILES C1C2(COC(O1)CCCN)COC(OC2)CCCN

Specifications

Melting Point 47°C
Color White
Boiling Point 204°C
Quantity 25 g
Formula Weight 274.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,9-Bis(3-aminopropyl)-2,4,8,10-tetraoxaspiro[5.5]undecane
EINECSNumber (5)-0843
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.