Learn More
Medchemexpress LLC 3,9-Dimethyl-3,9-diazaspiro[5.5]undecane | 93004-48-9 | 99.3% | C11H22N2 | 250 MG
SDP
Catalog No. 5000357643
Encompass_Preferred
3,9-Dimethyl-3,9-diazaspiro[5.5]undecane is a rigid PROTAC linker. It is utilized in the synthesis of PROTACs, such as FIP22, which is known as a potent and selective IRAK4 PROTAC degrader with anti-atopic dermatitis activity.
  • Rigid PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables the development of potent and selective IRAK4 PROTAC degraders
  • Supports research into treatments for inflammatory diseases
Catalog No. 50-003-57643 Supplier Medchemexpress LLC Supplier No. 250MGHY46871
3,9-Dimethyl-3,9-diazaspiro[5.5]undecane is a rigid PROTAC linker. It is utilized in the synthesis of PROTACs, such as FIP22, which is known as a potent and selective IRAK4 PROTAC degrader with anti-atopic dermatitis activity.
  • Rigid PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables the development of potent and selective IRAK4 PROTAC degraders
  • Supports research into treatments for inflammatory diseases