Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane

Catalog No. 1915235G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-152-35G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-152-35G Supplier Sigma Aldrich Supplier No. 1915235G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,9-Divinyl-2,4,8,10-tetraoxaspiro[5.5]undecane

Specifications

CAS 78-19-3
Melting Point 43°C to 46°C (lit.)
Density 1.251 g/mL (at 25°C (literature))
Boiling Point 108°C to 110°C (2 mmHg)
Molecular Formula C11H16O4
Linear Formula C11H16O4
MDL Number MFCD00006564
Quantity 500 mg
Molecular Weight (g/mol) 212.24
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.