Learn More
Echelon Biosciences Research Labs 3-A-AMINOCHOLESTANE10 mg
SDP
Supplier:  Echelon Biosciences Research Labs B034110MG
Encompass
SH2-Domain containing inositol 5-phosphatases (SHIP1 & SHIP2) dephosphorylate the 5-position of PI(3,4,5)P3 generating PI(3,4)P2. SHIP2 is ubiquitously expressed while SHIP1 is only found in hematopoietic lineage cells. 3AC is a selective inhibitor of SHIP1 (EC50 = 10 M) and shows no inhibition of the other isoform, SHIP2, at concentrations up to 1 mM. 3AC promotes apoptosis of SHIP1-expressing leukemia cells (KG-1) and multiple myeloma cells (OPM) suggesting SHIP1 inhibition is a potential drug target for blood cancers. Mice treated with 3AC show increased numbers of MIR cells in the spleen and lymph nodes and increased numbers of granulocytes. This item is non returnable
Catalog No. NC1475141