Learn More
AdipoGen 3-Acetamido-4-nitrophthalic an
SDP
Catalog No. 502561785
Encompass_Preferred
Chemical. CAS 209324-66-3. Formula C10H6N2O6. MW 250.17. Building Block/Intermediate used for the synthesis of diaminofluorescein derivatives.
Catalog No. 50-256-1785 Supplier AdipoGen Supplier No. CDXA0246G001
Chemical. CAS 209324-66-3. Formula C10H6N2O6. MW 250.17. Building Block/Intermediate used for the synthesis of diaminofluorescein derivatives.