Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetamidophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. A24535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2453-5G 5 g
A24531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2453-5G Supplier TCI America Supplier No. A24535G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 78887-39-5
Melting Point 140°C
Color Yellow
Molecular Formula C8H10BNO3
Quantity 5 g
MDL Number MFCD00236013
Synonym 3-acetylaminophenylboronic acid, 3-acetamidophenyl boronic acid, 3-acetamidobenzeneboronic acid, 3-acetylaminophenyl boronic acid, m-acetamidophenylboronic acid
InChI Key IBTSWKLSEOGJGJ-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)NC(=O)C)(O)O
IUPAC Name (3-acetamidophenyl)boronic acid
Molecular Weight (g/mol) 178.982
PubChem CID 157274
Formula Weight 178.98
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.