Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetamidopiperidine 99.0+%, TCI America™
SDP

Catalog No. A19345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19345G Supplier TCI America Supplier No. A19345G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5810-55-9
Molecular Formula C7H14N2O
Molecular Weight (g/mol) 142.202
InChI Key BCXSCZDWARFWAW-UHFFFAOYSA-N
PubChem CID 4169719
IUPAC Name N-piperidin-3-ylacetamide
SMILES CC(=O)NC1CCCNC1

Specifications

Melting Point 81°C
Color White-Yellow
Quantity 5 g
Formula Weight 142.20
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetamidopiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.