Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetamidopyrrolidine, TCI America™
SDP

Catalog No. A11085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A11085G Supplier TCI America Supplier No. A11085G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79286-74-1
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.175
MDL Number MFCD00059019
InChI Key HDCCJUCOIKLZNM-UHFFFAOYSA-N
PubChem CID 522715
IUPAC Name N-pyrrolidin-3-ylacetamide
SMILES CC(=O)NC1CCNC1

Specifications

Quantity 5 g
Formula Weight 128.18
Physical Form Liquid
Chemical Name or Material 3-Acetamidopyrrolidine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.