Learn More
STEMCELL Technologies 3% Acetic Acid with Methylene Blue, Size: 100 mL
SDP
Catalog No. NC9535453
Encompass
3% Acetic Acid with Methylene Blue is recommended for manual counting of nucleated mammalian cells. It will lyse red blood cell and white blood cell membranes. The remaining white blood cell nuclei will stain lightly with methylene blue.
Catalog No. NC9535453 Supplier STEMCELL Technologies Supplier No. 07060
3% Acetic Acid with Methylene Blue is recommended for manual counting of nucleated mammalian cells. It will lyse red blood cell and white blood cell membranes. The remaining white blood cell nuclei will stain lightly with methylene blue.