Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetoxy-1-propenylboronic acid pinacol ester, 97%

Catalog No. AAL1970003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1970003 1 g
AAL19700MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1970003 Supplier Thermo Scientific Chemicals Supplier No. L1970003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 161395-97-7 | C11H19BO4 | 226.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161395-97-7
Molecular Formula C11H19BO4
Molecular Weight (g/mol) 226.08
MDL Number MFCD03788751
InChI Key ZXBRLGZFCXGTBA-UHFFFAOYSA-N
Synonym 3-acetoxy-1-propenylboronic acid pinacol ester, e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl allyl acetate, 2-3-acetoxy-1-propenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-propen-1-yl acetate, 2e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl prop-2-en-1-yl acetate, e-3-acetoxy-1-propenylboronic acid 2,3-dimethylbutane-2,3-diyl ester
PubChem CID 11264613
IUPAC Name [(E)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)prop-2-enyl] acetate
SMILES CC(=O)OCC=CB1OC(C)(C)C(C)(C)O1

Specifications

Refractive Index 1.4485
Quantity 1 g
Beilstein 6923397
Formula Weight 226.08
Percent Purity 97%
Chemical Name or Material 3-Acetoxy-1-propenylboronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.