Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetoxybenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. A24675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24675G Supplier TCI America Supplier No. A24675G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34231-78-2
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00016603
InChI Key GVUMZPWBUAGJBP-UHFFFAOYSA-N
Synonym 3-acetoxybenzaldehyde, benzaldehyde, 3-acetyloxy, acetoxybenzaldehyde, acetic acid 3-formylphenyl ester, m-acetoxybenzaldehyde, m-ace-toxybenzaldehyde, meta-acetoxybenzaldehyde, pubchem13227, 3-formylphenyl acetate
PubChem CID 118618
IUPAC Name (3-formylphenyl) acetate
SMILES CC(=O)OC1=CC=CC(=C1)C=O

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 164.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Acetoxybenzaldehyde
EINECSNumber (3)-1185
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.