Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetyl-2-chloropyridine 98.0+%, TCI America™
SDP

Catalog No. A27955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27955G 5 g
A27951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27955G Supplier TCI America Supplier No. A27955G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55676-21-6
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD03840751
InChI Key WIWIOUAFBHZLNQ-UHFFFAOYSA-N
PubChem CID 10942697
IUPAC Name 1-(2-chloropyridin-3-yl)ethan-1-one
SMILES CC(=O)C1=C(Cl)N=CC=C1

Specifications

Boiling Point 114°C
Quantity 5 g
Formula Weight 155.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Acetyl-2-chloropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.