Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetyl-2-(methylaminosulfonyl)thiophene 98.0+%, TCI America™
SDP

Catalog No. A13155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A13155G Supplier TCI America Supplier No. A13155G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 138891-01-7
Molecular Formula C7H9NO3S2
Molecular Weight (g/mol) 219.273
MDL Number MFCD00191372
InChI Key OTWLSKTWKAMLMQ-UHFFFAOYSA-N
Synonym 3-Acetyl-2-thiophene-N-methylsulfonamide
PubChem CID 10632675
IUPAC Name 3-acetyl-N-methylthiophene-2-sulfonamide
SMILES CC(=O)C1=C(SC=C1)S(=O)(=O)NC

Specifications

Melting Point 105°C
Color White-Yellow
Quantity 5 g
Formula Weight 219.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetyl-2-(methylaminosulfonyl)thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.