Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetyl-5-bromopyridine, 97%

Catalog No. AAL1936203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1936203 1 g
AAL1936206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1936203 Supplier Thermo Scientific Chemicals Supplier No. L1936203
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 38940-62-4 | C7H6BrNO | 200.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38940-62-4
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.04
MDL Number MFCD03086033
InChI Key LDBPZEQZCOUYFT-UHFFFAOYSA-N
Synonym 3-acetyl-5-bromopyridine, 1-5-bromopyridin-3-yl ethanone, 1-5-bromopyridin-3-yl ethan-1-one, ethanone, 1-5-bromo-3-pyridinyl, 3-bromo-5-acetylpyridine, 1-5-bromo-3-pyridyl ethanone, 5-acetyl-3-bromopyridine, 1-5-bromo-3-pyridinyl ethanone, pubchem15626, acmc-1aglt
PubChem CID 820423
IUPAC Name 1-(5-bromopyridin-3-yl)ethanone
SMILES CC(=O)C1=CC(Br)=CN=C1

Specifications

Melting Point 77°C to 79°C
Quantity 1 g
Beilstein 112808
Formula Weight 200.03
Percent Purity 97%
Chemical Name or Material 3-Acetyl-5-bromopyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.