Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetyl-5-bromopyridine 97.0+%, TCI America™
SDP

Catalog No. A23765G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23765G 5 g
A23761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23765G Supplier TCI America Supplier No. A23765G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 38940-62-4
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.04
MDL Number MFCD03086033
InChI Key LDBPZEQZCOUYFT-UHFFFAOYSA-N
Synonym 3-acetyl-5-bromopyridine, 1-5-bromopyridin-3-yl ethanone, 1-5-bromopyridin-3-yl ethan-1-one, ethanone, 1-5-bromo-3-pyridinyl, 3-bromo-5-acetylpyridine, 1-5-bromo-3-pyridyl ethanone, 5-acetyl-3-bromopyridine, 1-5-bromo-3-pyridinyl ethanone, pubchem15626, acmc-1aglt
PubChem CID 820423
IUPAC Name 1-(5-bromopyridin-3-yl)ethan-1-one
SMILES CC(=O)C1=CC(Br)=CN=C1

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 5 g
Formula Weight 200.04
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetyl-5-bromopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.