Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetyl-6-bromocoumarin 96.0+%, TCI America™
SDP

Catalog No. A27235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27235G 5 g
A27231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27235G Supplier TCI America Supplier No. A27235G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2199-93-1
Molecular Formula C11H7BrO3
Molecular Weight (g/mol) 267.08
MDL Number MFCD00024075
InChI Key XFQYOFLFNKCHLG-UHFFFAOYSA-N
Synonym 3-acetyl-6-bromocoumarin, 3-acetyl-6-bromo-2h-chromen-2-one, 3-acetyl-6-bromo-chromen-2-one, 2h-1-benzopyran-2-one,3-acetyl-6-bromo, 3-acetyl-6-bromo-2-oxochromene, pubchem13336, maybridge1_000212, zerenex e/1034014, 6-bromanyl-3-ethanoyl-chromen-2-one
PubChem CID 304813
IUPAC Name 3-acetyl-6-bromo-2H-chromen-2-one
SMILES CC(=O)C1=CC2=CC(Br)=CC=C2OC1=O

Specifications

Melting Point 225°C
Quantity 5 g
Formula Weight 267.08
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Acetyl-6-bromocoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.