Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Acetyl-beta-boswellic acid | 5968-70-7 | 498.74 | 1 MG
SDP

Catalog No. 5000415259
Encompass_Preferred

3-Acetyl-beta-boswellic acid is a boswellic acid isolated from *Boswellia serrata* gum resin. It is for research use only and not sold to patients.

  • Purity: 98.10%
  • Appearance: Solid, white to off-white
  • Solubility: Soluble in DMSO (100 mg/mL)
  • Inhibits microsomal PGES1 in A549 cells with an IC50 of 3 μM
  • Shows cytotoxicity against human HL-60 cells with an IC50 of 10 μM after 48 hours
  • Shows cytotoxicity against human HeLa cells with an IC50 of 22 μM after 48 hours
  • Store at 4°C, sealed, away from moisture and light. In solvent, store at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-004-15259 Supplier Medchemexpress LLC Supplier No. HYN20751MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Acetyl-beta-boswellic acid is a boswellic acid isolated from *Boswellia serrata* gum resin. It is for research use only and not sold to patients.

  • Purity: 98.10%
  • Appearance: Solid, white to off-white
  • Solubility: Soluble in DMSO (100 mg/mL)
  • Inhibits microsomal PGES1 in A549 cells with an IC50 of 3 μM
  • Shows cytotoxicity against human HL-60 cells with an IC50 of 10 μM after 48 hours
  • Shows cytotoxicity against human HeLa cells with an IC50 of 22 μM after 48 hours
  • Store at 4°C, sealed, away from moisture and light. In solvent, store at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.