Learn More
Medchemexpress LLC 3-Acetyl-beta-boswellic acid | 5968-70-7 | MFCD03788773 | 498.74 | 5 MG
SDP
Catalog No. 5000416215
Encompass_Preferred
3-Acetyl-beta-boswellic acid is a boswellic acid isolated from Boswellia serrata gum resin. It is a pentacyclic triterpenoid compound.
  • Inhibits microsomal PGES1 in A549 cells (IC50: 3 μM).
  • Cytotoxic against human HL60 cells (IC50: 10 μM) and HeLa cells (IC50: 22 μM).
  • Shows antiproliferation activity against HUVEC cells (> 100 μM).
  • Inhibits human recombinant COX2 expressed in baculovirus infected Sf21 cells (> 40 μM).
Catalog No. 50-004-16215 Supplier Medchemexpress LLC Supplier No. HYN20755MG
3-Acetyl-beta-boswellic acid is a boswellic acid isolated from Boswellia serrata gum resin. It is a pentacyclic triterpenoid compound.
  • Inhibits microsomal PGES1 in A549 cells (IC50: 3 μM).
  • Cytotoxic against human HL60 cells (IC50: 10 μM) and HeLa cells (IC50: 22 μM).
  • Shows antiproliferation activity against HUVEC cells (> 100 μM).
  • Inhibits human recombinant COX2 expressed in baculovirus infected Sf21 cells (> 40 μM).