Learn More
Cayman Chemical 3-AcetylaconItIn 10mg
SDP
Catalog No. 502912694
Encompass_Preferred
A diterpene alkaloid with diverse biological activities; cytotoxic to P388 leukemia cells (IC50 = 18.6 µM); inhibits brevetoxin binding to (IC50 = 1.3 µM), and increases free sodium and calcium concentrations in, rat brain synaptosomes; reduces paw licking, flicking, and shaking in a mouse model of formalin-induced inflammatory pain at 100 UG/kg, i.v.
Catalog No. 50-291-2694 Supplier Cayman Chemical Supplier No. 3615510 MG
A diterpene alkaloid with diverse biological activities; cytotoxic to P388 leukemia cells (IC50 = 18.6 µM); inhibits brevetoxin binding to (IC50 = 1.3 µM), and increases free sodium and calcium concentrations in, rat brain synaptosomes; reduces paw licking, flicking, and shaking in a mouse model of formalin-induced inflammatory pain at 100 UG/kg, i.v.