Learn More
Medchemexpress LLC 3-Acetylcoumarin | 3949-36-8 | 97.0% | 188.18 | 1 ML
SDP
Catalog No. 5000350301
Encompass_Preferred
3-Acetylcoumarin (Compound 4) is a chemical compound demonstrating acaricidal activity against Tetranychus cinnabarinus. It also inhibits human recombinant MAO-B with an IC50 of 14.45 μM and exhibits cytotoxicity against human SH-SY5Y cells with an IC50 of 75 μM.
  • Exhibits acaricidal activity against Tetranychus cinnabarinus
  • Inhibits human recombinant MAO-B (IC50 of 14.45 μM)
  • Demonstrates cytotoxicity against human SH-SY5Y cells (IC50 of 75 μM)
Catalog No. 50-003-50301 Supplier Medchemexpress LLC Supplier No. HY13855410MM/1ML
3-Acetylcoumarin (Compound 4) is a chemical compound demonstrating acaricidal activity against Tetranychus cinnabarinus. It also inhibits human recombinant MAO-B with an IC50 of 14.45 μM and exhibits cytotoxicity against human SH-SY5Y cells with an IC50 of 75 μM.
  • Exhibits acaricidal activity against Tetranychus cinnabarinus
  • Inhibits human recombinant MAO-B (IC50 of 14.45 μM)
  • Demonstrates cytotoxicity against human SH-SY5Y cells (IC50 of 75 μM)