Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetylcoumarin, 98+%

Catalog No. aaa1253206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1253206 5 g
AAA1253214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1253206 Supplier Thermo Scientific Chemicals Supplier No. A1253206
Only null left
Add to Cart

CAS: 3949-36-8 | C11H8O3 | 188.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3949-36-8
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00006853
InChI Key CSPIFKKOBWYOEX-UHFFFAOYSA-N
Synonym 3-acetylcoumarin, 3-acetyl-2h-chromen-2-one, coumarin, 3-acetyl, 2h-1-benzopyran-2-one, 3-acetyl, 3-acetyl-2-oxochromene, 3-acetylcoumarine, pubchem8664, maybridge1_000885, prestwick0_000891, prestwick1_000891
PubChem CID 77553
IUPAC Name 3-acetylchromen-2-one
SMILES CC(=O)C1=CC2=CC=CC=C2OC1=O

Specifications

Melting Point 119°C to 124°C
Quantity 5 g
Beilstein 143498
Formula Weight 188.18
Percent Purity ≥98%
Chemical Name or Material 3-Acetylcoumarin
EINECSNumber 223-541-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.