Learn More
Cayman Chemical 3-Acetyldeoxy Nivalenol, Purity: ≥99%, CAS: 50722-38-8, 10 mg
SDP
Catalog No. 502951123
Encompass_Preferred
A mycotoxin; induces duodenal and splenic cell necrosis at 40 mg/kg, as well as lethality (LD50 = 70 mg/kg) in mice
Catalog No. 50-295-1123 Supplier Cayman Chemical Supplier No. 1142910 MG
A mycotoxin; induces duodenal and splenic cell necrosis at 40 mg/kg, as well as lethality (LD50 = 70 mg/kg) in mice