Learn More
Medchemexpress LLC 3-acetylindole | 703-80-0 | MFCD00005626 | 99.8% | 159.18 | C10H9NO | 100 G
SDP
Catalog No. 5000300846
Encompass_Preferred
3-Acetylindole is an indole-derived chemical intermediate commonly used in organic synthesis, notably as a precursor for antiviral agents and other heterocyclic compounds. It is supplied as a solid material with high reported purity for research and development use, and safety and analytical documentation are available.
Catalog No. 50-003-00846 Supplier Medchemexpress LLC Supplier No. HYW018667100G
3-Acetylindole is an indole-derived chemical intermediate commonly used in organic synthesis, notably as a precursor for antiviral agents and other heterocyclic compounds. It is supplied as a solid material with high reported purity for research and development use, and safety and analytical documentation are available.