Learn More
Medchemexpress LLC 3-acetylindole | 703-80-0 | MFCD00005626 | 99.8% | 159.18 | C10H9NO | 25 G
SDP
Catalog No. 5000302715
Encompass_Preferred
3-Acetylindole is an indole derivative supplied as a solid reagent used as a chemical synthesis precursor, notably for antiviral agent development and related small-molecule synthesis. It has the chemical formula C10H9NO, a molecular weight of 159.18, and a reported purity of 99.8%.
  • High reported purity suitable for synthesis applications.
  • Solid form convenient for storage and handling in organic synthesis.
  • Commonly used as a precursor in medicinal chemistry and antiviral research.
  • Soluble in common organic solvents, facilitating reaction setup and assays.
Catalog No. 50-003-02715 Supplier Medchemexpress LLC Supplier No. 25GHYW018667
3-Acetylindole is an indole derivative supplied as a solid reagent used as a chemical synthesis precursor, notably for antiviral agent development and related small-molecule synthesis. It has the chemical formula C10H9NO, a molecular weight of 159.18, and a reported purity of 99.8%.
  • High reported purity suitable for synthesis applications.
  • Solid form convenient for storage and handling in organic synthesis.
  • Commonly used as a precursor in medicinal chemistry and antiviral research.
  • Soluble in common organic solvents, facilitating reaction setup and assays.