Learn More
Cayman Chemical 3-AcetylmorphIn 10mg
SDP
Catalog No. 502893106
Encompass_Preferred
A less active metabolite of heroin with relatively weak affinity for µ-opioid receptors; intended only for forensic and research purposes
Catalog No. 50-289-3106 Supplier Cayman Chemical Supplier No. 900195410 MG
A less active metabolite of heroin with relatively weak affinity for µ-opioid receptors; intended only for forensic and research purposes