Learn More
Cayman Chemical 3-Acetylmorphine, Purity: ≥98%, CAS: 5140-28-3, 50 mg
SDP
Catalog No. 502893138
Encompass_Preferred
A less active metabolite of heroin with relatively weak affinity for µ-opioid receptors; intended only for forensic and research purposes
Catalog No. 50-289-3138 Supplier Cayman Chemical Supplier No. 900195450 MG
A less active metabolite of heroin with relatively weak affinity for µ-opioid receptors; intended only for forensic and research purposes