Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Acetylphenoxy)acetic Acid 98.0+%, TCI America™
SDP

Catalog No. A20615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20615G 5 g
A206125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20615G Supplier TCI America Supplier No. A20615G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1878-80-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD02180947
InChI Key VDZJGFDUSYCVJA-UHFFFAOYSA-N
PubChem CID 241887
IUPAC Name 2-(3-acetylphenoxy)acetic acid
SMILES CC(=O)C1=CC=CC(OCC(O)=O)=C1

Specifications

Melting Point 119°C
Color White
Quantity 5 g
Formula Weight 194.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (3-Acetylphenoxy)acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.