Learn More
Cayman Chemical 3-Acetylpyridine NAD, Purity: ≥95%, CAS: 86-08-8, 100 mg
SDP
Catalog No. 502995659
Encompass_Preferred
A NAD analog; used to study the mechanisms of oxidative phosphorylation
Catalog No. 50-299-5659 Supplier Cayman Chemical Supplier No. 21884100 MG
A NAD analog; used to study the mechanisms of oxidative phosphorylation