Learn More
Cayman Chemical 3-Acetylpyridine NAD, Purity: ≥95%, CAS: 86-08-8, 250 mg
SDP
Catalog No. 502986496
Encompass_Preferred
A NAD analog; used to study the mechanisms of oxidative phosphorylation
Catalog No. 50-298-6496 Supplier Cayman Chemical Supplier No. 21884250 MG
A NAD analog; used to study the mechanisms of oxidative phosphorylation